Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 14 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS115P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS115P 1 mL Unconjugated Concentrated 200μg/mL
MS115R7 7 mL Unconjugated Prediluted Prediluted
MS115B0 100 μL Biotin Concentrated 200μg/mL
MS115B1 500 μL Biotin Concentrated 200μg/mL
MS115B 1 mL Biotin Concentrated 200μg/mL
MS115P0 100 μL Unconjugated Concentrated 200μg/mL
MS115PABX 200 μL Unconjugated Concentrated 1mg/mL
MS115P1 500 μL Unconjugated Concentrated 200μg/mL
MS115R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS115P Supplier Epredia™ Supplier No. MS115P
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Mouse

Clone: LL002

Isotype: IgG3

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cytokeratin 14 Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LL002
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG3
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.