Learn More
Cytokeratin 14 Rabbit Polyclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 14, Rabbit Polyclonal Antibody.
Supplier: Epredia RB9020R1
Description
Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 14
Molecular Weight: 50 kDa
Positive Control: A431 cells; Skin or Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Cytokeratin 14 | |
Polyclonal | |
Unconjugated | |
A synthetic peptide derived from C-terminal of human Keratin 14 | |
IVD | |
Primary | |
Prediluted |
Western Blot | |
Polyclonal | |
Rabbit | |
1 mL | |
Cytoskeletal Protein | |
Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.