Learn More
Epredia™ Lab Vision™ Cytokeratin 15 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: LHK15
Isotype: IgG2a/κ
Species Reactivity: Human, Cow, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cytokeratin 15 Ab-1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | LHK15 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15 |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.