Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 15 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1068P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1068P1 500 μL 200μg/mL
MS1068P0 100 μL 200μg/mL
MS1068P 1 mL 200μg/mL
MS1068P1ABX 100 μL 1mg/mL
MS1068PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1068P1 Supplier Epredia™ Supplier No. MS1068P1
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunofluorescence applications with Epredia™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.

Keratin 15 is a type I keratin without a defined type II partner. Keratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail. Expression of keratin 15 is downregulated in some hyperproliferating situations, such as psoriasis and hypertrophic scars. Because keratinocytes in psoriasis and hypertrophic scars are activated, it is suggested that keratin 15 expression is not compatible with keratinocyte activation and the keratin 15 gene is downregulated to maintain the activated phenotype.

Host Species: Mouse

Clone: LHK15

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cytokeratin 15 Ab-1
Applications Immunofluorescence
Classification Monoclonal
Clone LHK15
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.