Learn More
Description
Host Species: Mouse
Clone: E3
Isotype: IgG2b/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Cytokeratin preparation from rat enterocytes
Molecular Weight: 46 kDa
Positive Control: Squamous cell CA, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin 17 Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | E3 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Keratin preparation from rat enterocytes |
| Purification Method | Squamous cell CA, skin |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.