Learn More
Description
Host Species: Mouse
Clone: DC10
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Human breast cancer PMC 42 cells
Molecular Weight: 45 kDa
Positive Control: HT29 cells, Skin, Breast or Lung CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin 18 Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | DC10 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Human breast cancer PMC 42 cells |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.