Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS142R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS142R1 1 mL Unconjugated Prediluted Prediluted
MS142P1 500 μL Unconjugated Concentrated 200μg/mL
MS142P 1 mL Unconjugated Concentrated 200μg/mL
MS142R7 7 mL Unconjugated Prediluted Prediluted
MS142B0 100 μL Biotin Concentrated 200μg/mL
MS142B1 500 μL Biotin Concentrated 200μg/mL
MS142B 1 mL Biotin Concentrated 200μg/mL
MS142P0 100 μL Unconjugated Concentrated 200μg/mL
MS142P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS142PABX 200 μL Unconjugated Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS142R1 Supplier Epredia Supplier No. MS142R1

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Keratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with keratin 8. It was reported that tissues from gastrointestinal tract are positive for both keratin 8 and 18 but do not contain keratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for keratin 18.

Host Species: Mouse

Clone: DC10

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Human breast cancer PMC 42 cells

Molecular Weight: 45 kDa

Positive Control: HT29 cells, Skin, Breast or Lung CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Specifications

Antigen Cytokeratin 18 Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DC10
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Human breast cancer PMC 42 cells
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.