Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 18 Rabbit Monoclonal Antibody

Catalog No. RM2108R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2108R1 1 mL Prediluted
RM2108S 1 mL Concentrated
RM2108S0 100 μL Concentrated
RM2108R7 7 mL Prediluted
RM2108RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2108R1 Supplier Epredia™ Supplier No. RM2108R1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 18, Rabbit Monoclonal Antibody.

Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Typically, it is absent in squamous cell carcinoma.

Host Species: Rabbit

Clone: E431-1

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues of the human CK18

Molecular Weight: 45 kDa

Positive Control: Mesothelioma, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E431-1
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues of the human CK18
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.