Learn More
Description
Host Species: Mouse
Clone: A53-B/A2.26; also known as Ks19.1
Isotype: IgG2a/λ
Species Reactivity: Human. Reacts weakly with Rat, Mouse and Guinea Pig. Others not known.
Epitope: aa 312-335
Immunogen: Human breast cancer MCF-7 cells
Molecular Weight: 40 kDa
Positive Control: HT29 and LS174T cells, skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin 19 Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | A53-B/A2.26; same as Ks19.1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human breast cancer MCF-7 cells |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.