Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 19 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS198PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Supernatant
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS198PABX 200 μL Concentrated 1mg/mL
MS198P1 500 μL Concentrated 200μg/mL
MS198P 1 mL Concentrated 200μg/mL
MS198P1ABX 100 μL Concentrated 1mg/mL
MS198R7 7 mL Prediluted
MS198R1 1 mL Prediluted
MS198RQ 12 mL Supernatant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS198PABX Supplier Epredia Supplier No. MS198PABX
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues, excluding the liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: A53-B/A2.26; also known as Ks19.1

Isotype: IgG2a/λ

Species Reactivity: Human. Reacts weakly with Rat, Mouse and Guinea Pig. Others not known.

Epitope: aa 312-335

Immunogen: Human breast cancer MCF-7 cells

Molecular Weight: 40 kDa

Positive Control: HT29 and LS174T cells, skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Specifications

Antigen Cytokeratin 19 Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A53-B/A2.26; same as Ks19.1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells
Quantity 200 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.