Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 19 Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1671P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1671P 1 mL Concentrated 200μg/mL
MS1671P1ABX 100 μL Concentrated 1mg/mL
MS1671PABX 200 μL Concentrated 1mg/mL
MS1671P1 500 μL Concentrated 200μg/mL
MS1671P0 100 μL Concentrated 200μg/mL
MS1671R1 1 mL Prediluted Prediluted
MS1671R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1671P Supplier Epredia Supplier No. MS1671P

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 19 Ab-4, Mouse Monoclonal Antibody.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: BA17

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Detergent soluble extract of human mammary epithelium

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cytokeratin 19 Ab-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone BA17
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Detergent soluble extract of human mammary epithelium
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.