Learn More
Description
Host Species: Mouse
Clone: BA17
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Detergent soluble extract of human mammary epithelium
Molecular Weight: 40 kDa
Positive Control: Colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cytokeratin 19 Ab-4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | BA17 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Detergent soluble extract of human mammary epithelium |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.