Learn More
Description
Host Species: Mouse
Clone: RCK108
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Cytoskeleton preparation of T24 cells
Molecular Weight: 40 kDa
Positive Control: LS174T and HT29 cells, Colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cytokeratin 19 Ab-5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | RCK108 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Cytoskeleton preparation of T24 cells |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.