Learn More
Epredia™ Lab Vision™ Cytokeratin 19 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 19
Molecular Weight: 40 kDa
Positive Control: LS174T
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cytokeratin 19 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from C-terminal of human Keratin 19 |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Cytoskeletal Protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.