Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 19 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9021R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9021R7 7 mL Prediluted
RB9021P1 500 μL Concentrated
RB9021P 1 mL Concentrated
RB9021R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB9021R7 Supplier Epredia™ Supplier No. RB9021R7

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Keratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 19

Molecular Weight: 40 kDa

Positive Control: LS174T

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cytokeratin 19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration Prediluted
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from C-terminal of human Keratin 19
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.