Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 5 Rabbit Monoclonal Antibody

Catalog No. RM2106RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2106RQ 12 mL Ready-to-Use
RM2106S 1 mL Concentrated
RM2106S0 100 μL Concentrated
RM2106R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM2106RQ Supplier Epredia™ Supplier No. RM2106RQ
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5, Rabbit Monoclonal Antibody.

Twenty human cytokeratins are divided into acidic (pI<5.7) and basic (pI>6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of cytokeratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of cytokeratins as markers in cancer research and tumor identification. Point mutations in cytokeratin-5 gene may cause Epidermolysis Bullosa Simplex and Dowling-Degos disease. cytokeratin-5 is expressed in most epithelioid and biphasic mesotheliomas, thymoma, squamous and other carcinomas. It is related to cytokeratin-6.

Host Species: Rabbit

Clone: EP1601Y

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK5

Molecular Weight: 59 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP1601Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues on the C-term of human CK5
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.