Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1352R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1352R1 1 mL Prediluted Prediluted
MS1352P 1 mL Concentrated 200μg/mL
MS1352RQ 12 mL Ready-to-Use Prediluted
MS1352P1ABX 100 μL Concentrated 1mg/mL
MS1352PABX 200 μL Concentrated 1mg/mL
MS1352P1 500 μL Concentrated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1352R1 Supplier Epredia Supplier No. MS1352R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.

Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).

Host Species: Mouse

Clone: OV-TL 12/30

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: OTN II ovarian carcinoma cell line

Molecular Weight: 54 kDa

Positive Control: Ovarian CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 7 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OV-TL 12/30
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen OTN II ovarian carcinoma cell line
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.