Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 8 Rabbit Monoclonal Antibody

Catalog No. RM2107RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2107RQ 12 mL Ready-to-Use
RM2107S 1 mL Concentrated
RM2107R7 7 mL Prediluted
RM2107S0 100 μL Concentrated
RM2107R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2107RQ Supplier Epredia™ Supplier No. RM2107RQ

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8, Rabbit Monoclonal Antibody.

Cytokeratin 8 belongs to the type B (basic) subfamily of low molecular weight keratins and usually exists in combination with cytokeratin 18. Cytokeratin 8 is primarily found in the non-squamous simple epithelia. It is expressed in mesothelioma and most adenocarcinomas of simple epithelium including breast ducts, gall bladder, intestine, liver, pancreas and prostate.

Host Species: Rabbit

Clone: EP1628Y

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK8

Molecular Weight: 52 kDa

Positive Control: Pancreas, Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP1628Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues of the human CK18
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.