Learn More
Epredia™ Lab Vision™ DNA-PKcs Ab-2 Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.
Host Species: Mouse
Clone: 25-4
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: aa 3198-4127
Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells
Molecular Weight: 460 kDa
Positive Control: LS174T cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Gel Supershift (Not optimum)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | DNA-PKcs Ab-2 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 25-4 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human DNA-dependent Protein Kinase purified from HeLa cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.