Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ DNA-PKcs Ab-2 Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS370PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS370PABX 200 μL 1mg/mL
MS370P0 100 μL 200μg/mL
MS370P1 500 μL 200μg/mL
MS370P 1 mL 200μg/mL
MS370P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS370PABX Supplier Epredia™ Supplier No. MS370PABX

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 25-4

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: aa 3198-4127

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift (Not optimum)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen DNA-PKcs Ab-2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 25-4
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Quantity 200 μL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.