Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ DNA-PKcs Ab-4 (Cocktail) Mouse Monoclonal Antibody, 1mg/mL

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.

Supplier:  Epredia™ MS423PABX

Catalog No. MS423PABX


Description

Description

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2 + 25-4 + 42-psc

Isotype: IgG1 + IgG2a + IgG1

Species Reactivity: Human, Mouse, Hamster, Xenopus and Chicken. Others not known.

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil or testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

DNA-PKcs Ab-4 (Cocktail)
Monoclonal
1mg/mL
Mouse
200 μL
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
18-2 + 25-4 + 42-psc
Unconjugated
Human DNA-dependent Protein Kinase purified from HeLa cells
RUO
Chicken, Hamster, Human, Mouse
IgG1, IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.