Learn More
Description
Host Species: Mouse
Clone: 1.1
Isotype: Unknown
Species Reactivity: Human. Others not known.
Immunogen: Synthetic peptide of human DOG1
Molecular Weight: 78 kDa
Positive Control: Gastrointestinal Stromal Tumors
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Dog1 |
| Applications | Immunohistochemistry |
| Clone | 1.1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Synthetic peptide of human Dog1 |
| Quantity | 12 mL |
| Primary or Secondary | Primary |
| Form | Ready-to-Use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.