Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ DOG1 Rabbit Monoclonal Antibody

Catalog No. RM9132S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9132S0 100 μL Concentrated
RM9132R1 1 mL Prediluted
RM9132R7 7 mL Prediluted
RM9132RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM9132S0 Supplier Epredia™ Supplier No. RM9132S0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Dog1, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

DOG1 is a cell surface protein of unknown function selectively expressed in GIST. Among GIST cases with KIT mutations the DOG1 antibody identified 11% more cases than c-Kit antibody. DOG1 identifies vast majority of both c-Kit negative and PDGFRA mutated GIST cases that may still benefit from imatinib mesylate (Gleevac), an inhibitor of the kit tyrosin kinase. In addition, DOG1 immunoreactivity is seen in fewer cases of mesanchymal, epithelial tumors and melanomas when compared with c-Kit. The use of this highly sensitive and specific novel marker will increase the accuracy of GIST diagnosis.

Host Species: Rabbit

Clone: SP31

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Synthetic peptide of human DOG1

Molecular Weight: 78 kDa

Positive Control: Gastrointestinal Stromal Tumors

Cellular Localization: Cytoplasmic and Membranous

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen DOG1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP31
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide of human Dog1
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.