Learn More
Epredia™ Lab Vision™ DP-1 Transcription Factor Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 1DP06 (TFD10)
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 83-204
Immunogen: Recombinant DP-1 protein
Molecular Weight: 45 kDa
Positive Control: MAD109 or Raji cells, Tonsil
Cellular Localization: Cytoplasm and nucleus
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | DP-1 Transcription Factor Ab-6 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1DP06; same as TFD10 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant DP-1 protein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.