Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ DP-1 Transcription Factor Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1056R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1056R1 1 mL
MS1056P0 100 μL 200μg/mL
MS1056P1 500 μL 200μg/mL
MS1056P 1 mL 200μg/mL
MS1056P1ABX 100 μL 1mg/mL
MS1056PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1056R1 Supplier Epredia™ Supplier No. MS1056R1

Ensure accurate, reproducible results in western blotting experiments with Epredia™ DP-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.

DP family contains at least two members, DP-1 and -2. Structurally, DP-1 and -2 are relatively conserved, yet their expression appears to be tissue specific. E2F-1, a functional target for the tumor suppressor protein Rb, heterodimers with DP-1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.

Host Species: Mouse

Clone: 1DP06 (TFD10)

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 83-204

Immunogen: Recombinant DP-1 protein

Molecular Weight: 45 kDa

Positive Control: MAD109 or Raji cells, Tonsil

Cellular Localization: Cytoplasm and nucleus

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen DP-1 Transcription Factor Ab-6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1DP06; same as TFD10
Concentration
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant DP-1 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.