Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ E Cadherin Mouse Monoclonal Antibody with BSA and azide

Catalog No. MS9470P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS9470P1 500 μL Concentrated
MS9470P0 100 μL Concentrated
MS9470P 1 mL Concentrated
MS9470R1 1 mL Prediluted
MS9470RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS9470P1 Supplier Epredia™ Supplier No. MS9470P1

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ Cadherin E, Mouse Monoclonal Antibody.

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Host Species: Mouse

Clone: SPM471

Isotype: IgG2b/κ

Species Reactivity: Human. Others not tested.

Epitope: 600-707aa

Immunogen: Recombinant protein encoding human Cadherin-E 600-707aa

Molecular Weight: 120kDa

Positive Control: Colon CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cadherin E/E-Cadherin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SPM471
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding human Cadherin-E 600-707aa
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.