Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ E-Cadherin Rabbit Monoclonal Antibody

Catalog No. RM2100S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2100S0 100 μL Concentrated
RM2100S 1 mL Concentrated
RM2100R7 7 mL Prediluted
RM2100RQ 12 mL Ready-to-Use
RM2100R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2100S0 Supplier Epredia™ Supplier No. RM2100S0
Add to Cart
Add to Cart

Ensure accurate and reliable results in immunohistochemistry applications with Epredia™ Cadherin E, Rabbit Monoclonal Antibody.

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominantly in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis. E-cadherin is used in distinguishing LCIS from DCIS and that of ILC from IDC in indeterminate cases.

Host Species: Rabbit

Clone: EP700Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from the fifth cadherin domain of human cadherin

Molecular Weight: 135/80kDa

Positive Control: Breast Carcinoma

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen E-Cadherin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP700Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from the fifth cadherin domain of human cadherin
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.