Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)

Catalog No. p-4532329 Shop All Epredia Products
Click to view available options
Quantity:
125mL
500mL

Catalog No. AP9004125

Supplier: Epredia™ AP9004125

Improve the reactivity of many antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X).

  • 10mM EDTA buffer, pH 8.0
  • Dilute 10-fold with distilled water before use

Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C

Specifications

Content And Storage Room temperature
Dilution 10X
Product Type EDTA
Quantity 125mL
For Use With (Application) Immunohistochemistry
pH 8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.