Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-10 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS378B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS378B1 500 μL Biotin Concentrated 200μg/mL
MS378B0 100 μL Biotin Concentrated 200μg/mL
MS378P1 500 μL Unconjugated Concentrated 200μg/mL
MS378P0 100 μL Unconjugated Concentrated 200μg/mL
MS378P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS378PABX 200 μL Unconjugated Concentrated 1mg/mL
MS378R1 1 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS378B1 Supplier Epredia Supplier No. MS378B1

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 111.6

Isotype: IgG1

Species Reactivity: Human. Not mouse and rat.

Epitope: Extracellular domain

Immunogen: Extracellular domain of human recombinant EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells; Skin, Placenta or Squamous cell CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)
  • Effect on EGF Binding (Inhibitory)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-10
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 111.6
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Extracellular domain of human recombinant EGFR protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.