Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-13 Mouse Monoclonal Antibody

Catalog No. MS609PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS609PABX 200 μL Unconjugated 1mg/mL
MS609B0 100 μL Biotin 200μg/mL
MS609B1 500 μL Biotin 200μg/mL
MS609B 1 mL Biotin 200μg/mL
MS609P0 100 μL Unconjugated 200μg/mL
MS609P 1 mL Unconjugated 200μg/mL
MS609P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS609PABX Supplier Epredia™ Supplier No. MS609PABX

Obtain accurate, reproducible results in immunoprecipitation experiments with Epredia™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528 + 199.12

Isotype: IgG2a + IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-13
Applications Immunoprecipitation
Classification Monoclonal
Clone 528 + 199.12
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)
Quantity 200 μL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.