Learn More
Epidermal Growth Factor Receptor/EGFR Ab-2 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 225
Isotype: IgG1
Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.
Epitope: Extracellular domain
Immunogen: Purified EGFR from A431 cells
Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)
Positive Control: A431 cells
Cellular Localization: Membrane
Recommended for:
- Immunoprecipitation (Native only)
- Blocks EGF/TGFα-Induced Activation of EGFR
- Arrests Tumor Growth in vivo
- Inhibits Binding of Ab-1 (clone 528) to EGFR
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Epidermal Growth Factor Receptor/EGFR Ab-2 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 225 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified EGFR from A431 cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.