Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial Membrane Antigen/EMA Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS741R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS741R7 7 mL Prediluted
MS741P0 100 μL Concentrated 200μg/mL
MS741P1 500 μL Concentrated 200μg/mL
MS741P 1 mL Concentrated 200μg/mL
MS741P1ABX 100 μL Concentrated 1mg/mL
MS741PABX 200 μL Concentrated 1mg/mL
MS741R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS741R7 Supplier Epredia Supplier No. MS741R7
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-3, Mouse Monoclonal Antibody.

Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.

Host Species: Mouse

Clone: E29

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: RPAP

Immunogen: Delipidated extract of human milk fat globule membranes

Molecular Weight: 265-400 kDa

Positive Control: Breast or Colon CA

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Epithelial Membrane Antigen/EMA Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E29
Concentration
Conjugate Unconjugated
Host Species Mouse
Immunogen Delipidated extract of human milk fat globule membranes
Quantity 7 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.