Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody

Catalog No. MS1898R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1898R7 7 mL Prediluted
MS1898S0 100 μL Concentrated
MS1898S 1 mL Concentrated
MS1898R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1898R7 Supplier Epredia™ Supplier No. MS1898R7
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: Ber-EP4

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: MCF-7 human breast carcinoma cell line

Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ber-EP4
Conjugate Unconjugated
Host Species Mouse
Immunogen MCF-7 human breast carcinoma cell line
Quantity 7 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.