Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody

Catalog No. MS1458R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1458R7 7 mL Prediluted
MS1458S1 500 μL Concentrated
MS1458S0 100 μL Concentrated
MS1458S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1458R7 Supplier Epredia™ Supplier No. MS1458R7A

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epstein-Barr Virus/LMP Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: CS1 + CS2 + CS3 + CS4

Isotype: IgG1/κ

Species Reactivity: EBV-positive cells

Epitope: Located in the internal part of the membrane-associated viral LMP

Immunogen: EBV-encoded recombinant latent membrane protein

Molecular Weight: 60 kDa (LMP)

Positive Control: EBV-positive lymphoblastoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Epstein-Barr Virus/LMP Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CS1+CS2+CS3+CS4
Conjugate Unconjugated
Host Species Mouse
Immunogen EBV-encoded recombinant latent membrane protein
Quantity 7 mL
Regulatory Status ASR, IVD
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Ebola virus
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.