Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Estrogen Receptor Ab-10 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS315P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS315P1 500 μL Unconjugated 200μg/mL
MS315B0 100 μL Biotin 200μg/mL
MS315B1 500 μL Biotin 200μg/mL
MS315P 1 mL Unconjugated 200μg/mL
MS315P1ABX 100 μL Unconjugated 1mg/mL
MS315PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS315P1 Supplier Epredia™ Supplier No. MS315P1

Ensure accurate, reproducible results in western blotting, immunoprecipitation, gel supershift and chromatin precipitation experiments.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: TE111.5D11

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Dog, Mouse, Rat, Sheep, Rabbit, Hamster and Chicken. Others not known.

Epitope: aa 302-553

Immunogen: 35 kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli

Molecular Weight: 67 kDa

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift
  • Blocks Binding of Estrogen to ER
  • Chromatin Precipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Estrogen Receptor Ab-10
Applications Immunoprecipitation
Classification Monoclonal
Clone TE111.5D11
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen 35kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli
Quantity 500 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.