Learn More
Epredia™ Lab Vision™ Estrogen Receptor Ab-10 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: TE111.5D11
Isotype: IgG1
Species Reactivity: Human, Cow, Pig, Dog, Mouse, Rat, Sheep, Rabbit, Hamster and Chicken. Others not known.
Epitope: aa 302-553
Immunogen: 35 kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli
Molecular Weight: 67 kDa
Positive Control: T47D cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunoprecipitation (Native and denatured)
- Gel Supershift
- Blocks Binding of Estrogen to ER
- Chromatin Precipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Estrogen Receptor Ab-10 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | TE111.5D11 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | 35kDa, C-terminal fragment (aa 302-595) of human ER expressed in E. coli |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.