Learn More
Description
The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Hamster. Does not react with rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of human estrogen receptor, α protein
Molecular Weight: 67 kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
- Gel Supershift (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Estrogen Receptor Ab-16 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified w/o BSA and Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from the C-terminus of human estrogen receptor, alpha protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.