Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein
Molecular Weight: 67 kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Estrogen Receptor Ab-17 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein |
| Purification Method | Purified with BSA and azide |
| Quantity | 500 μL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.