Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Estrogen Receptor Ab-21 Rat Monoclonal Antibody with BSA and azide

Catalog No. RT1641PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200µg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
RT1641PABX 200 μL 1mg/mL
RT1641P1 500 μL 200µg/mL
RT1641P 1 mL 200µg/mL
RT1641P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RT1641PABX Supplier Epredia™ Supplier No. RT1641PABX

Ensure accurate, reproducible results in western blotting and ELISA experiments with Epredia™ Estrogen Receptor Ab-21, Rat Monoclonal Antibody.

ER protein exhibits six functionally discrete domains required for transcription activation, binding to estrogen response element (ERE), constitutive dimerization, binding to heat shock proteins, and ligand recognition. It is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors may improve response to anti-estrogen therapy.

Host Species: Rat

Clone: H222

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Shows broad species reactivity.

Immunogen: Estrogen receptor purified from MCF-7 cells

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Estrogen Receptor Ab-21
Applications Western Blot
Classification Monoclonal
Clone H222
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Estrogen receptor purified from MCF-7 cells
Quantity 200 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.