Learn More
Description
Host Species: Rabbit
Clone: SP1
Isotype: IgG
Species Reactivity: Human, Mouse and Hamster. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide derived from C-terminal of human estrogen receptor
Molecular Weight: 67 kDa
Positive Control: Breast CA, MCF-7 cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence (Not verified)
- Flow Cytometery (Not verified)
Specifications
Specifications
| Antigen | Estrogen Receptor (SP1) |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | SP1 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from C-terminal of human estrogen receptor |
| Quantity | 7 mL |
| Regulatory Status | IVD |
| Research Discipline | Steroids Hormones and Growth Factors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.