Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor (SP1) Rabbit Monoclonal Antibody, Epredia™

Catalog No. RM9101S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Conjugate:
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form
RM9101S 1 mL Unconjugated Concentrated
RM9101S0 100 μL Unconjugated Concentrated
RM9101S1 500 μL Unconjugated Concentrated
RM9101R7 7 mL Unconjugated Prediluted
RM9101RQ 12 mL Unconjugated Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. RM9101S Supplier Epredia Supplier No. RM9101S
This item is not returnable. View return policy

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Estrogen Receptor (SP1), Rabbit Monoclonal Antibody.

Recognizes a protein of 67 kDa, which is identified as estrogen receptor (ER) alpha. The ER gene consists of more than 140kb of genomic DNA divided into 8 exons, and is translated into a protein with six functionally discrete domains, labeled A through F. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Clone: SP1

Isotype: IgG

Species Reactivity: Human, Mouse and Hamster. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide derived from C-terminal of human estrogen receptor

Molecular Weight: 67 kDa

Positive Control: Breast CA, MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence (Not verified)
  • Flow Cytometery (Not verified)

Specifications

Antigen Estrogen Receptor (SP1)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP1
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminal of human estrogen receptor
Quantity 1 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Hamster, Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.