Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Ezrin/p81/80K/Cytovillin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS661R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS661R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS661R1 Supplier Epredia™ Supplier No. MS661R1

Specifically detect ezrin in human, monkey, cow, kangaroo, rat, mouse and hamster samples with Epredia™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody.

Ezrin (also known as p81, 80K, or cytovillin) is a cytoplasmic protein enriched in microvilli and other cell surface structures. Its NH2-terminal domain is associated with the plasma membrane and the COOH-terminal domain with the cytoskeleton. Ezrin has an actin-binding capability. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. Ezrin serves as a tyrosine kinase substrate, and is phosphorylated in EGF-stimulated cells.

Host Species: Mouse

Clone: 3C12

Isotype: IgG1

Species Reactivity: Human, Monkey, Cow, Kangaroo, Rat, Mouse and Hamster. Others not known.

Epitope: aa 362-585

Immunogen: A recombinant fragment of human ezrin corresponding to aa 362-585

Molecular Weight: 81 kDa

Positive Control: Raji cells, Lung

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Ezrin/p81/80K/Cytovillin Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3C12
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen A recombinant fragment of human ezrin corresponding to aa 362-585
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Bovine, Hamster, Human, Kangaroo, Mouse, Non-human Primate, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.