Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ α-Fetoprotein (AFP) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS226P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
Concentration:
1mg/mL
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS226P1 500 μL 200μg/mL
MS226P1ABX 100 μL 1mg/mL
MS226PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS226P1 Supplier Epredia™ Supplier No. MS226P1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ α-Fetoprotein (AFP) Ab-1, Mouse Monoclonal Antibody.

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Mouse

Clone: C3

Isotype: IgG2a

Species Reactivity: Human, Dog and Pig. It does not react with cow, cat, mouse and rat.

Epitope: Not determined

Immunogen: Raised against affinity purified α-fetoprotein (AFP) from serum of a hepatoma patient

Molecular Weight: 70 kDa

Positive Control: Fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen alpha-Fetoprotein (AFP) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C3
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Raised against affinity purified alpha fetoprotein (AFP) from serum of a hepatoma patient
Quantity 500 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Canine, Human, Pig
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.