Learn More
Epredia™ Lab Vision™ α-Fetoprotein (AFP) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: C3
Isotype: IgG2a
Species Reactivity: Human, Dog and Pig. It does not react with cow, cat, mouse and rat.
Epitope: Not determined
Immunogen: Raised against affinity purified α-fetoprotein (AFP) from serum of a hepatoma patient
Molecular Weight: 70 kDa
Positive Control: Fetal liver or hepatocellular CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | alpha-Fetoprotein (AFP) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | C3 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Raised against affinity purified alpha fetoprotein (AFP) from serum of a hepatoma patient |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.