Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Fibronectin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS165P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
Concentration:
1mg/mL
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS165P 1 mL 200μg/mL
MS165P0 100 μL 200μg/mL
MS165PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS165P Supplier Epredia™ Supplier No. MS165P

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and biological blockade experiments.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: HFN 7.1

Isotype: IgG1

Species Reactivity: Human specific

Epitope: Peptide core

Immunogen: Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Positive Control: SW156 cells

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade (Order Ab without azide)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Fibronectin Ab-1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HFN 7.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Quantity 1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.