Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filaggrin Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS449R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS449R7 7 mL Prediluted -
MS449P1ABX 100 μL Concentrated 1mg/mL
MS449PABX 200 μL Concentrated 1mg/mL
MS449P1 500 μL Concentrated 200μg/mL
MS449P0 100 μL Concentrated 200μg/mL
MS449P 1 mL Concentrated 200μg/mL
MS449R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS449R7 Supplier Epredia Supplier No. MS449R7

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Filaggrin Ab-1, Mouse Monoclonal Antibody.

Filaggrin is expressed only in well-differentiated, keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.

Host Species: Mouse

Clone: FLG01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant filaggrin protein

Molecular Weight: 38 kDa (Filaggrin) and 400 kDa (Profilaggrin)

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Filaggrin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FLG01
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant filaggrin protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.