Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-1/VEGFR1 Ab-1 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB1527P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
RB1527P0 100 μL Purified with BSA and Azide
RB1527P1 500 μL Purified with BSA and Azide
RB1527P1ABX 500 μL Purified w/o BSA and Azide
RB1527P 1 mL Purified with BSA and Azide
RB1527PABX 1 mL Purified w/o BSA and Azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB1527P0 Supplier Epredia Supplier No. RB1527P0

Ensure accurate, reproducible results in western blotting experiments with Epredia™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody.

Flt-1/vascular endothelial growth factor receptor 1 (VEGFR1), a cell membrane receptor kinase, is a high-affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flk-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin

Molecular Weight: 180 kDa

Positive Control: Hey cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Flt-1/VEGFR1 Ab-1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified with BSA and Azide
Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.