Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin
Molecular Weight: 180 kDa
Positive Control: Hey cells
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Flt-1/VEGFR1 Ab-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified w/o BSA and Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.