Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1449P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1449P1ABX 100 μL Concentrated 1mg/mL
MS1449PABX 200 μL Concentrated 1mg/mL
MS1449R1 1 mL Prediluted -
MS1449R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1449P1ABX Supplier Epredia Supplier No. MS1449P1ABX

Epredia™ Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

FSH is a pituitary hormone involvrd in the maturation of ovarian follicles and estrogen secretion in females. In the pituitary gland, FSH is produced by gonadotrophs In males, FSH stimulates the secretion of testosterone.

Host Species: Mouse

Clone: FSH03

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Immunogen: Purified human FSH-β

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Follicle Stimulating Hormone (FSH) Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FSH03
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human FSH-Beta
Quantity 100 μL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Bovine, Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.