Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-1 (SPM498) Mouse Monoclonal Antibody, Epredia™

Catalog No. p-4546367 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS10637R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS10637R1 Supplier Epredia Supplier No. MS10637R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ GLUT-1, Mouse Monoclonal Antibody.

Glucose is fundamental to the metabolism in mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.

Host Species: Mouse

Clone: SPM498

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1

Molecular Weight: 55 kDa

Positive Control: Esophageal CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen GLUT-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM498
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide derived from C-terminal of human GLUT-1
Purification Method Esophageal CA
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.